EMAIL THIS PAGE TO A FRIEND
Cerebellum (London, England)
Cerebellar Changes in Guinea Pig Offspring Following Suppression of Neurosteroid Synthesis During Late Gestation.
PMID 27255705
Abstract
Elevated gestational concentrations of allopregnanolone are essential for the development and neuroprotection of the foetal brain. Preterm birth deprives the foetus of these high levels of allopregnanolone, which may contribute to the associated adverse effects on cerebellar development. Preterm birth alters expression of GABA