EMAIL THIS PAGE TO A FRIEND
Oncotarget
Folate-targeted star-shaped cationic copolymer co-delivering docetaxel and MMP-9 siRNA for nasopharyngeal carcinoma therapy.
PMID 27259274
Abstract
The co-delivery of drug and gene has become the primary strategy in cancer therapy. Based on our previous work, to co-deliver docetaxel (DOC) and MMP-9 siRNA more efficiently for HNE-1 nasopharyngeal carcinoma therapy, a folate-modified star-shaped copolymer (FA-CD-PLLD) consisting of β-cyclodextrin (CD) and poly(L-lysine) dendron (PLLD) was synthesized, and then used for DOC and MMP-9 co-delivery. Different from commonly used amphiphilic copolymers micelles, the obtained CD derivative could be used directly for the combinatorial delivery of nucleic acid and hydrophobic DOC without a complicated micellization process. In vitro and in vivo assays are carried out to confirm the effectiveness of the target strategy and combined treatment. It was found that the conjugation of CD-PLLD with FA could enhance the DOC/MMP-9 delivery effect obviously, inducing a more significant apoptosis and decreasing invasive capacity of HEN-1 cells. In vivo assays showed that FA-CD-PLLD/DOC/MMP-9 could inhibit HNE-1 tumor growth and decrease PCNA expression effectively, indicating a promising strategy for nasopharyngeal carcinoma therapy. Moreover, the in vivo distribution of DOC and MMP-9, blood compatibility and toxicity are also explored.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU010531 MISSION® esiRNA, esiRNA targeting human MMP9 (esiRNA1)