EMAIL THIS PAGE TO A FRIEND
Allergology international : official journal of the Japanese Society of Allergology
A panel study of airborne particulate matter composition versus concentration: Potential for inflammatory response and impaired pulmonary function in children.
PMID 27262618
Abstract
The relationship between airborne particulate matter (PM) and pulmonary function in children has not been consistent among studies, potentially owing to differences in the inflammatory response to PM, based on PM types and sources. The objective of this study was to investigate the effect of airborne PM on pulmonary function in schoolchildren and its potential for an inflammatory response. Daily morning peak expiratory flow (PEF) was measured in 339 schoolchildren in February 2015. Interleukin (IL)-8 production was assessed in THP1 cells stimulated by airborne PM collected every day during the study period, and these IL-8 concentrations are described as the daily IL-8 levels. A linear mixed model was used to estimate the association between PEF values and the daily levels of suspended PM (SPM), PM diameters smaller than 2.5 μm (PM The daily IL-8 levels were significantly associated with those of SPM and PM These findings suggest that the effects of airborne PM on pulmonary function in schoolchildren might depend more on the pro-inflammatory response than the mass concentration of the PM.