EMAIL THIS PAGE TO A FRIEND
Luminescence : the journal of biological and chemical luminescence
Second bioluminescence-activating component in the luminous fungus Mycena chlorophos.
PMID 27271205
Abstract
Mycena chlorophos is an oxygen-dependent bioluminescent fungus. The mechanisms underlying its light emission are unknown. A component that increased the bioluminescence intensity of the immature living gills of M. chlorophos was isolated from mature M. chlorophos gills and chemically characterized. The bioluminescence-activating component was found to be trans-3,4-dihydroxycinnamic acid and its bioluminescence activation was highly structure-specific.