EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Allicin inhibits oxidative stress-induced mitochondrial dysfunction and apoptosis by promoting PI3K/AKT and CREB/ERK signaling in osteoblast cells.
PMID 27284348
Abstract
Osteoporosis is a disease of the skeleton that is characterized by the loss of bone mass and degeneration of bone microstructure, resulting in an increased risk of fracture. Oxidative stress, which is known to promote oxidative damage to mitochondrial function and also cell apoptosis, has been recently indicated to be implicated in osteoporosis. However, there are few agents that counteract oxidative stress in osteoporosis. In the present study, the protective effects of allicin against the oxidative stress-induced mitochondrial dysfunction and apoptosis were investigated in murine osteoblast-like MC3T3-E1 cells. The results demonstrated that allicin counteracted the reduction of cell viability and induction of apoptosis caused by hydrogen peroxide (H
Related Materials