EMAIL THIS PAGE TO A FRIEND
Artificial organs
Multimodal Imaging for In Vivo Evaluation of Induced Pluripotent Stem Cells in a Murine Model of Heart Failure.
PMID 27296083
Abstract
Myocardial stem cell therapy in heart failure is strongly dependent on successful cellular transfer, engraftment, and survival. Moreover, massive cell loss directly after intramyocardial injection is commonly observed, generating the need for efficient longitudinal monitoring of transplanted cells in order to develop more efficient transplantation techniques. Therefore, the aim of the present study was to assess viability and cardiac retention of induced pluripotent stem cells after intramyocardial delivery using in vivo bioluminescence analysis (BLI) and magnetic resonance imaging (MRI). Murine induced pluripotent stem cells (iPSCs) were transfected for luciferase reporter gene expression and labeled intracellularly with supraparamagnetic iron oxide particles. Consequently, 5 × 10