EMAIL THIS PAGE TO A FRIEND
Journal of veterinary science
Physical-chemical and biological characterization of different preparations of equine chorionic gonadotropin.
PMID 27297410
Abstract
Ovarian stimulation with commercial preparations of equine chorionic gonadotropin (eCG) produces extremely variable responses in domestic animals, ranging from excessive stimulation to practically no stimulation, when applied on the basis of their declared unitage. This study was conducted to analyze four commercial preparations from different manufacturers via reversed-phase HPLC (RP-HPLC) in comparison with a reference preparation and an official International Standard from the World Health Organization. The peaks obtained by this qualitative and quantitative physical-chemical analysis were compared using an