EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Ecotoxicity evaluation of a WWTP effluent treated by solar photo-Fenton at neutral pH in a raceway pond reactor.
PMID 27335017
Abstract
Some pollutants can be resistant to wastewater treatment, hence becoming a risk to aquatic and terrestrial biota even at the very low concentrations (ng L
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
655546
1,6-Bis(p-carboxyphenoxy)hexane, 90%
C20H22O6
D6875
21-Hydroxyprogesterone, ≥97% (HPLC)
C21H30O3