EMAIL THIS PAGE TO A FRIEND
Drug development and industrial pharmacy
Solubility and dissolution performances of spray-dried solid dispersion of Efavirenz in Soluplus.
PMID 27349377
Abstract
Efavirenz (EFV), a first-line anti-HIV drug largely used as part of antiretroviral therapies, is practically insoluble in water and belongs to BCS class II (low solubility/high permeability). The aim of this study was to improve the solubility and dissolution performances of EFV by formulating an amorphous solid dispersion of the drug in polyvinyl caprolactam-polyvinyl acetate-polyethylene glycol graft copolymer (Soluplus
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2706
Ammonium acetate solution, for molecular biology, 7.5 M
C2H7NO2