EMAIL THIS PAGE TO A FRIEND
Experimental animals
Generation of a transgenic mouse line for conditional expression of human IL-6.
PMID 27349442
Abstract
IL-6 is a cytokine that is involved in various physiological and pathological conditions, and approaches using gain-of-function transgenic animals have contributed in elucidating IL-6 function. However, studies of the multiple functions of IL-6 in vivo are very time consuming because they require the generation of transgenic mice that harbor the gene encoding IL-6 under the control of specific promoters to mimic different pathologies. Here, we report the establishment of a conditional human IL-6 transgenic mouse, LGL-IL6, which conditionally expresses human IL-6 by taking advantage of the well-characterized Cre recombinase drivers.