EMAIL THIS PAGE TO A FRIEND
Stem cells (Dayton, Ohio)
Human Mesenchymal Stem Cell-Derived Microvesicles Prevent the Rupture of Intracranial Aneurysm in Part by Suppression of Mast Cell Activation via a PGE2-Dependent Mechanism.
PMID 27350036
Abstract
Activation of mast cells participates in the chronic inflammation associated with cerebral arteries in intracranial aneurysm formation and rupture. Several studies have shown that the anti-inflammatory effect of mesenchymal stem cells (MSCs) is beneficial for the treatment of aneurysms. However, some long-term safety concerns exist regarding stem cell-based therapy for clinical use. We investigated the therapeutic potential of microvesicles (MVs) derived from human MSCs, anuclear membrane bound fragments with reparative properties, in preventing the rupture of intracranial aneurysm in mice, particularly in the effect of MVs on mast cell activation. Intracranial aneurysm was induced in C57BL/6 mice by the combination of systemic hypertension and intrathecal elastase injection. Intravenous administration of MSC-derived MVs on day 6 and day 9 after aneurysm induction significantly reduced the aneurysmal rupture rate, which was associated with reduced number of activated mast cells in the brain. A23187-induced activation of both primary cultures of murine mast cells and a human mast cell line, LAD2, was suppressed by MVs treatment, leading to a decrease in cytokine release and tryptase and chymase activities. Upregulation of prostaglandin E2 (PGE2) production and E-prostanoid 4 (EP4) receptor expression were also observed on mast cells with MVs treatment. Administration of an EP4 antagonist with the MVs eliminated the protective effect of MVs against the aneurysmal rupture in vivo. Human MSC-derived MVs prevented the rupture of intracranial aneurysm, in part due to their anti-inflammatory effect on mast cells, which was mediated by PGE2 production and EP4 activation. Stem Cells 2016;34:2943-2955.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7388
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide
C30H36N6O9