EMAIL THIS PAGE TO A FRIEND
Environmental toxicology
Microcystin-LR induces a wide variety of biochemical changes in the A549 human non-small cell lung cancer cell line: Roles for protein phosphatase 2A and its substrates.
PMID 27352821
Abstract
Our previous studies have described the toxic effects of microcystin-LR (MC-LR) in various normal cell lines and human hepatoma SMMC-7721 cells, but the specific effects of MC-LR in other types of cancer cells with respect to protein phosphatase 2A (PP2A) have not been fully elaborated. A549 human lung adenocarcinoma cells have been identified to express organic anion-transporting polypeptides (OATP) involved in cellular uptake of MC-LR, and thus probably make an appropriate in vitro model to assess MC-LR's cytotoxicity. Hence, in our present study, A549 cells were treated with various concentrations of MC-LR for 24 h. The presence of MC-LR in A549 cells was confirmed, and PP2A activity, PP2A substrates, cytoskeleton, apoptosis, and proliferation were subsequently explored. The results showed that 5-10 μM MC-LR inhibited PP2A activity significantly but 0.5-1 μM MC-LR did not change PP2A activity dramatically. The inhibition could result from the hyperphosphorylation of PP2A/C at Tyr307, an elevation in the total PP2A/C expression and the dissociation of α4/PP2A/C complexes. Moreover, MC-LR led to rearrangements of filamentous actin and microtubules, which might be correlated with the hyperphosphorylation of Ezrin, VASP and HSP27 due to PP2A inhibition and mitogen-activated protein kinase (MAPK) activation. However, exposure to MC-LR for 24 h failed to trigger either apoptosis or proliferation, which might be related to PP2A-inhibition-induced hyperphosphorylation of Bcl-2 and Bad and the activation status of Akt. In conclusion, our data indicated that MC-LR induced extensive molecular and cellular alterations in A549 cells through a PP2A-centered pathway, which differed in some respects from our previous study in SMMC-7721 cells. To our knowledge, this is the first report comprehensively demonstrating the effects of MC-LR in A549 cells, and our findings provide insights into the mechanism of MC-LR toxicity in cancer cells. © 2016 Wiley Periodicals, Inc. Environ Toxicol 32: 1065-1078, 2017.
Related Materials