EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Association of thiocolchicine with tubulin.
PMID 2735908
Abstract
Thiocolchicine, a colchicine analog in which the C-10 methoxy is replaced with a thiomethyl moiety, was shown to bind with high affinity to the colchicine site on tubulin (Ka = 1.07 +/- 0.14 x 10(6) M-1 at 23 degrees C). Like colchicine, the association kinetics were biphasic, and the rate constants of both phases were temperature dependent. The rate constant of the fast phase of the association was 4 times greater than the rate constant for colchicine binding, and the activation energy was lower (19.1 +/- 1.8 kcal/mol). X-ray crystallographic analysis shows that thiocolchicine displays greater puckering of the tropone C ring than colchicine (Koerntgen, C. and Margulis, T. N. (1977) J. Pharm. Sci. 66, 1127-1131.). These results indicate that the conformation of the C ring may have little effect on the energetics of colchicinoids binding to tubulin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0954 Thiocolchicine, ≥97% (HPLC)
C22H25NO5S