EMAIL THIS PAGE TO A FRIEND
Cell research
Identification of blood vascular endothelial stem cells by the expression of protein C receptor.
PMID 27364685
Abstract
Vascular growth and remodeling are dependent on the generation of new endothelial cells from stem cells and the involvement of perivascular cells to maintain vessel integrity and function. The existence and cellular identity of vascular endothelial stem cells (VESCs) remain unclear. The perivascular pericytes in adult tissues are thought to arise from the recruitment and differentiation of mesenchymal progenitors during early development. In this study, we identified Protein C receptor-expressing (Procr