EMAIL THIS PAGE TO A FRIEND
The Analyst
Integrated on-chip mass spectrometry reaction monitoring in microfluidic devices containing porous polymer monolithic columns.
PMID 27373801
Abstract
Chip-based microfluidics enable the seamless integration of different functions into single devices. Here, we present microfluidic chips containing porous polymer monolithic columns as a means to facilitate chemical transformations as well as both downstream chromatographic separation and mass spectrometric analysis. Rapid liquid phase lithography prototyping creates the multifunctional device economically.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4500
Nα-Benzoyl-L-arginine ethyl ester hydrochloride, trypsin substrate
C15H22N4O3 · HCl