EMAIL THIS PAGE TO A FRIEND
Molecular carcinogenesis
CD24 is a genetic modifier for risk and progression of prostate cancer.
PMID 27377469
Abstract
CD24 plays an oncogenic role in the onset and progression of various human cancers, including prostate cancer. In the present study, we identified two linkage disequilibrium blocks with four recombination hotspot motifs in human CD24 locus. To elucidate whether genetic variants of CD24 are associated with susceptibility to prostate cancer and its disease status, we conducted a case-control association study with two P170 C/T and P-534 A/C polymorphisms of CD24 in 590 patients with prostate cancer and 590 healthy controls. A significant increased risk of prostate cancer was found in men with the P170
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T25003
Tetranitromethane
CN4O8