EMAIL THIS PAGE TO A FRIEND
Bioinformatics (Oxford, England)
Recognizing metal and acid radical ion-binding sites by integrating ab initio modeling with template-based transferals.
PMID 27378301
Abstract
More than half of proteins require binding of metal and acid radical ions for their structure and function. Identification of the ion-binding locations is important for understanding the biological functions of proteins. Due to the small size and high versatility of the metal and acid radical ions, however, computational prediction of their binding sites remains difficult. We proposed a new ligand-specific approach devoted to the binding site prediction of 13 metal ions (Zn http://zhanglab.ccmb.med.umich.edu/IonCom CONTACT: hxz@imut.edu.cn or zhng@umich.eduSupplementary information: Supplementary data are available at Bioinformatics online.