EMAIL THIS PAGE TO A FRIEND
Journal of magnetic resonance imaging : JMRI
Delayed gadolinium-enhanced MRI of cartilage (dGEMRIC) and T
PMID 27381489
Abstract
To assess the feasibility of delayed gadolinium-enhanced magnetic resonance imaging (MRI) of cartilage (dGEMRIC) and T Seventeen patients with wrist pain (mean age, 41.4 ± 13.1 years) including a subgroup with chondromalacia (n = 11) and 15 healthy volunteers (26.0 ± 2.2 years) underwent dGEMRIC and T The optimum time window was 45-90 minutes, and the 60-minute timepoint was subsequently used. In chondromalacia, dGEMRIC values were lower (551 ± 84 msec, P < 0.001), and T At 3T, dGEMRIC and T 3 J. Magn. Reson. Imaging 2017;45:381-389.