EMAIL THIS PAGE TO A FRIEND
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas
Role of adenylyl cyclase in reduced β-adrenoceptor-mediated vasorelaxation during maturation.
PMID 27383122
Abstract
Beta-adrenergic receptor (βAR)-dependent blood vessel relaxation is impaired in older animals and G protein activation has been suggested as the causative mechanism. Here, we investigated the role of βAR subtypes (β1AR, β2AR, and β3AR) and cAMP in maturation-dependent vasorelaxation impairment. Aortic rings from 15 Sprague-Dawley male rats (3 or 9 weeks old) were harvested and left intact or denuded of the endothelium. Vascular relaxation in aortic rings from younger and older groups was compared in the presence of βAR subtype agonists and antagonists along with cAMP and cGMP antagonists. Isolated aortic rings were used to evaluate relaxation responses, protein expression was evaluated by western blot or real time PCR, and metabolites were measured by ELISA. Expression of βAR subtypes and adenylyl cyclase was assessed, and cAMP activity was measured in vascular tissue from both groups. Isoproterenol- and BRL744-dependent relaxation in aortic rings with and without endothelium from 9-week-old rats was impaired compared with younger rats. The β1AR antagonist CGP20712A (10-7 M) did not affect isoproterenol or BRL744-dependent relaxation in arteries from either group. The β2AR antagonist ICI-118,551 (10-7 M) inhibited isoproterenol-dependent aortic relaxation in both groups. The β3AR antagonist SR59230A (10-7 M) inhibited isoproterenol- and BRL744-dependent aortic ring relaxation in younger but not in older rats. All βAR subtypes were expressed in both groups, although β3AR expression was lower in the older group. Adenylyl cyclase (SQ 22536) or protein kinase A (H89) inhibitors prevented isoproterenol-induced relaxation in younger but not in older rats. Production of cAMP was reduced in the older group. Adenylyl cyclase III and RyR3 protein expression was higher in the younger group. In conclusion, altered expression of β3AR and adenylyl cyclase III may be responsible for reduced cAMP production in the older group.