EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
High-Efficiency Nonfullerene Polymer Solar Cells with Medium Bandgap Polymer Donor and Narrow Bandgap Organic Semiconductor Acceptor.
PMID 27386802
Abstract
A nonfullerene polymer solar cell with a high efficiency of 9.26% is realized by using benzodithiophene-alt-fluorobenzotriazole copolymer J51 as a medium-bandgap polymer donor and the low-bandgap organic semiconductor ITIC with high extinction coefficients as the acceptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
901058
J51