EMAIL THIS PAGE TO A FRIEND
Chemical biology & drug design
Docetaxel-Chitosan nanoparticles for breast cancer treatment: cell viability and gene expression study.
PMID 27390258
Abstract
Docetaxel acts through the inhibition of tubulin polymerization and reduction in the expression of BCL-2 gene. In this study, nanoparticles containing Docetaxel were prepared and their effects on the gene expression levels of BCL-2 and BAX genes were investigated. The drug was first conjugated to chitosan, and the nanoparticles were assembled in the presence of hyaluronic acid. Conjugations were confirmed by