EMAIL THIS PAGE TO A FRIEND
Sangyo igaku. Japanese journal of industrial health
[Method for simultaneous determination of six metabolites of toluene, xylene and ethylbenzene, and its application to exposure monitoring of workers in a printing factory with gravure machines].
PMID 2739101
Abstract
For the biological monitoring of exposure to solvent composed of toluene, xylene, and ethylbenzene used in a printing factory with gravure machines, we developed a HPLC method for the simultaneous determination of urinary metabolites of this solvent, i.e. hippuric acid, o-, m-, and p-methylhippuric acid, mandelic acid and phenylglyoxylic acid. Except for phenylglyoxylic acid, urinary concentrations of the metabolites determined by the present method correlated well with the air concentrations of the respective solvent components. Hence the present method is useful in monitoring solvent exposure. In 91 workers of the printing factory and 53 control subjects, we also determined the concentrations of some phenolic metabolites and confirmed that o-cresol is a useful indicator for monitoring toluene exposure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B13055
Phenylglyoxylic acid, 97%
C8H6O3
78610
Phenylglyoxylic acid, purum, ≥98.0% (T)
C8H6O3