EMAIL THIS PAGE TO A FRIEND
Oncotarget
Long non-coding RNA SPRY4-IT1 pormotes colorectal cancer metastasis by regulate epithelial-mesenchymal transition.
PMID 27391336
Abstract
Colorectal cancer (CRC) remains one of the most common cancers worldwide. Increasing evidence indicates that SPRY4 intronic transcript 1 (SPRY4-IT1) regulate cell growth, differentiation, apoptosis, and cancer progression. However, the expression and function of SPRY4-IT1 in the progression of CRC remains largely unknown. Here, we reported that SPRY4-IT1 was upregulated in CRC. Increased SPRY4-IT1 expression in CRC was associated with larger tumor size and higher clinical stage. In vitro experiments revealed that SPRY4-IT1 knockdown significantly inhibited CRC cell proliferation by causing G1 arrest and promoting apoptosis, whereas SPRY4-IT1 overexpression promoted cell proliferation. Further functional assays indicated that SPRY4-IT1 overexpression significantly promoted cell migration and invasion by regulate the epithelial-mesenchymal transition (EMT). Taken together, our study demonstrates that SPRY4-IT1 could act as a functional oncogene in CRC, as well as a potential therapeutic target to inhibit CRC metastasis.