EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Thermal stability and decompositions kinetics under non-isothermal conditions of imatinib mesylate α form.
PMID 27392171
Abstract
The thermal decomposition and kinetic parameters of synthetized imatinib mesylate α form α form were determined by thermogravimetry (TGA/DTG) under non-isothermal conditions. The experiments were performed at a 25-940°C temperature range at five different heating rates: 2.5Kmin(-1), 5Kmin(-1), 10Kmin(-1), 15Kmin(-1) and 20Kmin(-1) per minute in a nitrogen atmosphere. Imatinib mesylate α form presents one-step mass loss during the degradation process. The thermal stability of the examined material, the melting temperature (Tonset=220.6°C) and ΔH fusion=-95.74Jg(-1) at a heating rate of 10°Cmin(-1) was established. The values of activation energies have been estimated using Kissinger, Flynn-Wall-Ozawa (FWO) and Kissinger-Akahira-Sunose (KAS) methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS022173
Imatinib
C29H31N7O