Biological trace element research
Changes in Trace Element Contents and Morphology in Bones of Duck Exposed to Molybdenum or/and Cadmium.
PMID 27392954
Cadmium (Cd) and high molybdenum (Mo) can lead to adverse reactions on animals, but the coinduced toxicity of Mo and Cd to bone in ducks was not well understood. The objective of this study was to investigate the changes in trace elements' contents and morphology in bones of duck exposed to Mo or/and Cd. One hundred twenty healthy 11-day-old male ducks were randomly divided into six groups and treated with commercial diet containing Cd or/and Mo. On the 60th and 120th days, the blood, excretion, and metatarsals were collected to determine alkaline phosphatase (ALP) activity and the contents of Mo, Cd, calcium (Ca), phosphorus (P), copper (Cu), iron (Fe), zine (Zn), and selenium (Se). In addition, metatarsals were subjected to histopathological analysis with the optical microscope and radiography. The results indicated that Mo and Cd contents significantly increased while Ca, P, Cu, and Se contents remarkably decreased in metatarsals in coexposure groups (Pxa0xa00.05). Ca content in serum had no significant difference among experimental groups (Pxa0>xa00.05), but P content was significantly decreased in HMo and HMoxa0+xa0Cd groups (Pxa0