EMAIL THIS PAGE TO A FRIEND
ESC heart failure
Chronic hypoxia down-regulates tight junction protein ZO-2 expression in children with cyanotic congenital heart defect.
PMID 27398226
Abstract
Tight junction protein zonula occludens protein 2 (ZO-2) is a member of the membrane-associated guanylate kinases protein family known to be expressed at tight junctions of epithelial and endothelial cells and at adherens junctions (AJs) in cardiomyocytes. Little is known about ZO-2 expression and function in the human heart. Here, we examined the hypothesis that chronic hypoxia down-regulates ZO-2 expression in human myocardium and cultured rat cardiomyocytes. Patients with a diagnosis of cyanotic ( Hypoxia down-regulates ZO-2 expression in both cyanotic patient's myocardium and cultured rat cardiomyocytes. This down-regulation suggest an involvement of ZO-2 in cardiac remodelling of AJs in cyanotic children and may explain the greater susceptibility of cyanotic patients to corrective heart surgery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2206
Anti-β-Catenin antibody produced in rabbit, whole antiserum