EMAIL THIS PAGE TO A FRIEND
Journal of biomedical materials research. Part B, Applied biomaterials
Magnesium release from mesoporous carriers on endosseus implants does not influence bone maturation at 6 weeks in rabbit bone.
PMID 27405685
Abstract
The release of magnesium ions (Mg Titanium implants were coated with mesoporous titania layers and either loaded with Mg Mg The results suggested that the initial enhancement in osseointegration induced by magnesium release from mesoporous titania coatings has no detrimental effects during bone maturation. © 2016 Wiley Periodicals, Inc. J Biomed Mater Res Part B: Appl Biomater, 105B: 2118-2125, 2017.