EMAIL THIS PAGE TO A FRIEND
BioFactors (Oxford, England)
Methyl gallate, a potent antioxidant inhibits mouse and human adipocyte differentiation and oxidative stress in adipocytes through impairment of mitotic clonal expansion.
PMID 27412172
Abstract
Methyl gallate (MG) is a derivative of gallic acid and a potent antioxidant. In this study, we confirmed that MG treatment effectively inhibits lipid accumulation, which occurred mostly in the early stages of adipogenesis. We also showed that shortly after adipogenic induction, MG facilitated a G0/G1 cell cycle arrest. Mechanistic studies revealed that MG treatment inhibited ERK1/2 phosphorylation, which is a key regulator of the G1- to S-phase transition. Furthermore, MG treatment prevented the adipogenic hormonal stimuli-induced inhibition of the cyclin-dependent kinase inhibitor p27
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274194
Methyl 3,4,5-trihydroxybenzoate, 98%
C8H8O5