EMAIL THIS PAGE TO A FRIEND
European journal of clinical nutrition
Hematological complications in anorexia nervosa.
PMID 27436150
Abstract
Anemia, leukopenia and, although less frequently, thrombocytopenia are possible hematological complications of anorexia nervosa considered strictly secondary to chronic malnutrition. This is a retrospective study on the prevalence of these disorders in a large cohort of 318 female patients with AN (20.4±5.6 years, body mass index (BMI) 15.9±1.6 kg/m Patients were studied on an outpatient basis after obtaining medical history, clinical examination, routine hematobiochemical and endocrine tests, electrocardiography, psychiatric interview and bioelectrical impedance analysis and, in particular, phase angle determination. All patients with other comorbidities, in particular with mean corpuscular volume <80 fl, were excluded for suspected genetic alteration in the synthesis of hemoglobin. Hematologic data showed that 16.7% of patients had anemia, 7.9% neutropenia and 8.9% thrombocytopenia. These abnormalities were strictly related to the duration of illness (P=0.028), and to protein energy malnutrition, in particular, BMI and phase angle (P<0.001). Our study offers description of the incidence of hematologic defects in a selected and large sample of AN female patients, suggesting that its incidence is related to the degree and duration of protein energy malnutrition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0531
BIO-Acetoxime, ≥98% (HPLC)
C18H12BrN3O3