EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Substantiation of maximum permissible levels of antimony trioxide and pentasulide in the atmospheric air of inhabitated places].
PMID 2744535
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11133
Antimony(V) sulfide, technical, ≥60% Sb basis
S5Sb2