EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Polynuclear genital cells in rat spermatogenesis in the normal state and after exposure to xenobiotics].
PMID 2744563
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45475
Ethiofencarb, PESTANAL®, analytical standard
C11H15NO2S