EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Spatial distribution of triazine residues in a shallow alluvial aquifer linked to groundwater residence time.
PMID 27448811
Abstract
At present, some triazine herbicides occurrence in European groundwater, 13 years after their use ban in the European Union, remains of great concern and raises the question of their persistence in groundwater systems due to several factors such as storage and remobilization from soil and unsaturated zone, limited or absence of degradation, sorption in saturated zones, or to continuing illegal applications. In order to address this problem and to determine triazine distribution in the saturated zone, their occurrence is investigated in the light of the aquifer hydrodynamic on the basis of a geochemical approach using groundwater dating tracers (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34054
Simazine-d10, PESTANAL®, analytical standard
C7H2D10ClN5