EMAIL THIS PAGE TO A FRIEND
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP
Comprehensive characterization of three glutathione S-transferase family proteins from black rockfish (Sebastes schlegelii).
PMID 27449269
Abstract
Glutathione S-transferases (GSTs, EC 2.5.1.18) are categorized as phase II enzymes, which form an important multifunctional family associated with a wide variety of catalytic activities. GSTω, GSTρ, and GSTθ are cytosolic GSTs which have been extensively studied in a variety of organisms; however, few studies have focused on teleosts. Those paralogs from black rockfish (Sebastes schlegelii; RfGSTω, RfGSTρ, and RfGSTθ, respectively) were molecularly, biochemically, and functionally characterized to determine their antioxidant extent and protective aptitudes upon pathogenic stress. RfGSTω, RfGSTρ, and RfGSTθ, contained open reading frames of 717bp, 678bp, and 720bp respectively, which encoded respective proteins of 239, 226, and 240 amino acids in length. In silico analysis revealed that all RfGSTs possessed characteristic N-terminal domains bearing glutathione (GSH)-binding sites, and C-terminal domains containing substrate-binding sites. Recombinant RfGSTω (rRfGSTω) catalyzed the conjugation of GSH to dehydroascorbate (DHA), while rRfGSTθ and rRfGSTρ catalyzed to the model GST substrate 1-Chloro-2,4-dinitrobenzene (CDNB). Kinetic analysis revealed variation in Km and Vmax values for each rRfGST, indicating their different conjugation rates. The optimum conditions (pH and temperature) and inhibition assays of each protein demonstrated different optimal ranges showing their wide range of activity as an assembly. RfGSTω and RfGSTθ paralogs demonstrated their antioxidant potential towards H2O2 and heavy metals (Cd, Zn, and Cu) in vitro, while RfGSTρ had an antioxidant potential only towards heavy metals (Zn and Cu). Though all the paralogs were ubiquitously expressed in different magnitudes, RfGSTω was highly expressed in blood, whereas RfGSTρ and RfGSTθ were highly expressed in liver. The mRNA expression of RfGSTω and RfGSTθ, upon Streptococcus iniae and poly I:C stimulation, revealed a significantly up-regulated expression, whereas RfGSTρ mRNA expression was significantly down-regulated. Collectively, our findings suggest that RfGSTω, RfGSTρ, and RfGSTθ paralogs are potent in detoxifying xenobiotic toxics, capable of protecting cells from oxidative stress generated by both H2O2 and heavy metals, and finally, yet importantly, stimulated under pathogenic stress signals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3281
Hematin porcine
C34H33FeN4O5