EMAIL THIS PAGE TO A FRIEND
Human vaccines & immunotherapeutics
The prophylactic effects of human IgG derived from sera containing high anti-PcrV titers against pneumonia-causing Pseudomonas aeruginosa.
PMID 27454613
Abstract
The PcrV cap structure of the type III secretory apparatus of Pseudomonas aeruginosa is a vaccine target. Human immunoglobulin G (IgG) molecules extracted from sera containing high or low anti-PcrV titers were tested for their effects against P. aeruginosa pneumonia in a mouse model. Among 198 volunteers, we selected the top 10 high anti-PcrV titer sera and the bottom 10 low anti-PcrV titer sera and extracted the IgG fraction from each serum sample. First, we examined the effects of the IgG against virulent P. aeruginosa. A lethal dose of P. aeruginosa premixed with saline, low titer human IgG, high titer human IgG, or rabbit-derived polyclonal anti-PcrV IgG was intratracheally administered into the lungs of mice, and their survival and lung inflammation were evaluated for 24 h. The high anti-PcrV titer human IgG had a prophylactic effect. Next, the prophylactic effects of intravenous administration of extracted and pooled high or low anti-PcrV titer human IgG were examined. Here, prophylactic intravenous administration of pooled high anti-PcrV titer human IgG, which showed binding capacity to P. aeruginosa PcrV, was more effective than the administration of its low titer pooled equivalent, and the measured physiological and inflammatory parameters correlated with the anti-PcrV titer levels. This result indirectly implies that high anti-PcrV titers in blood can help to protect against virulent P. aeruginosa infections. In addition, the IgG fractions from such high titer sera have potential to be a source of specific intravenous immunoglobulin products for passive vaccination against virulent P. aeruginosa infections.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9416
TWEEN® 20, for molecular biology, viscous liquid