EMAIL THIS PAGE TO A FRIEND
Applied microbiology and biotechnology
A binary concrete crack self-healing system containing oxygen-releasing tablet and bacteria and its Ca
PMID 27459869
Abstract
A strategy to supply molecular oxygen for microbial calcium precipitation was developed for the first time. Firstly, a controlled oxygen-releasing tablet (ORT) containing CaO