EMAIL THIS PAGE TO A FRIEND
Drug testing and analysis
Determination of 21 drugs in oral fluid using fully automated supported liquid extraction and UHPLC-MS/MS.
PMID 27464485
Abstract
Collection of oral fluid (OF) is easy and non-invasive compared to the collection of urine and blood, and interest in OF for drug screening and diagnostic purposes is increasing. A high-throughput ultra-high-performance liquid chromatography-tandem mass spectrometry method for determination of 21 drugs in OF using fully automated 96-well plate supported liquid extraction for sample preparation is presented. The method contains a selection of classic drugs of abuse, including amphetamines, cocaine, cannabis, opioids, and benzodiazepines. The method was fully validated for 200 μL OF/buffer mix using an Intercept OF sampling kit; validation included linearity, sensitivity, precision, accuracy, extraction recovery, matrix effects, stability, and carry-over. Inter-assay precision (RSD) and accuracy (relative error) were <15% and 13 to 5%, respectively, for all compounds at concentrations equal to or higher than the lower limit of quantification. Extraction recoveries were between 58 and 76% (RSD < 8%), except for tetrahydrocannabinol and three 7-amino benzodiazepine metabolites with recoveries between 23 and 33% (RSD between 51 and 52 % and 11 and 25%, respectively). Ion enhancement or ion suppression effects were observed for a few compounds; however, to a large degree they were compensated for by the internal standards used. Deuterium-labelled and