EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Zinc Induces Dendritic Cell Tolerogenic Phenotype and Skews Regulatory T Cell-Th17 Balance.
PMID 27465530
Abstract
Zinc (Zn) is an essential metal for development and maintenance of both the innate and adaptive compartments of the immune system. Zn homeostasis impacts maturation of dendritic cells (DCs) that are important in shaping T cell responses. The mechanisms by which Zn regulates the tolerogenic phenotype of DCs remain largely unknown. In this study, we investigated the effect of Zn on DC phenotype and the generation of Foxp3(+) regulatory T cells (Tregs) using a model of Histoplasma capsulatum fungal infection. Exposure of bone marrow-derived DCs to Zn in vitro induced a tolerogenic phenotype by diminishing surface MHC class II (MHCII) and promoting the tolerogenic markers, programmed death-ligand (PD-L)1, PD-L2, and the tryptophan degrading enzyme, IDO. Zn triggered tryptophan degradation by IDO and kynurenine production by DCs and strongly suppressed the proinflammatory response to stimulation by TLR ligands. In vivo, Zn supplementation and subsequent H. capsulatum infection supressed MHCII on DCs, enhanced PD-L1 and PD-L2 expression on MHCII(lo) DCs, and skewed the Treg-Th17 balance in favor of Foxp3(+) Tregs while decreasing Th17 cells. Thus, Zn shapes the tolerogenic potential of DCs in vitro and in vivo and promotes Tregs during fungal infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
447439
1-Methyl-L-tryptophan, 95%
C12H14N2O2