EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and chemistry
Effects of carbon dioxide on juveniles of the freshwater mussel (Lampsilis siliquoidea [Unionidae]).
PMID 27466973
Abstract
Carbon dioxide (CO