EMAIL THIS PAGE TO A FRIEND
Cell death and differentiation
IL-12p40 impairs mesenchymal stem cell-mediated bone regeneration via CD4
PMID 27472064
Abstract
Severe or prolonged inflammatory response caused by infection or biomaterials leads to delayed healing or bone repair failure. This study investigated the important roles of the proinflammatory cytokines of the interleukin-12 (IL-12) family, namely, IL-12 and IL-23, in the inflammation-mediated inhibition of bone formation in vivo. IL-12p40
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0605
Z-Asp(O-Me)-Glu(O-Me)-Val-Asp(O-Me) fluoromethyl ketone, ≥90% (HPLC)
C30H41N4O12F