EMAIL THIS PAGE TO A FRIEND
European journal of cancer prevention : the official journal of the European Cancer Prevention Organisation (ECP)
Grape skin extract mitigates tissue degeneration, genotoxicity, and oxidative status in multiple organs of rats exposed to cadmium.
PMID 27472085
Abstract
The aim of this study was to investigate whether grape skin extract can mitigate the noxious activities induced by cadmium exposure in multiple organs of rats. For this purpose, histopathological analysis for the liver, genotoxicity, and oxidative status in the blood and liver were investigated in this setting. A total of 20 Wistar rats weighing 250 g, on average, and 8 weeks of age were distributed into four groups (n=5) as follows: control group (nontreated group); cadmium group (Cd); and grape skin extract groups (Cd+GS) at 175 or 350 mg/l. Histopathological analysis in liver showed that animals treated with grape skin extract showed improved tissue degeneration induced by cadmium intoxication. Genetic damage was reduced in blood and hepatocytes as indicated by comet and micronucleus assays in animals treated with grape skin extract. Copper-zinc superoxide dismutase and cytochrome c gene expression increased in groups treated with grape skin extract in liver cells. Grape skin extract also reduced the 8-hydroxy-2'-deoxyguanosine levels in liver cells compared with the cadmium group. Taken together, our results indicate that grape skin extract can mitigate tissue degeneration, genotoxicity, and oxidative stress induced by cadmium exposure in multiple organs of Wistar rats.