EMAIL THIS PAGE TO A FRIEND
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology
Molecular characterisation and antifungal susceptibility of clinical Cryptococcus deuterogattii (AFLP6/VGII) isolates from Southern Brazil.
PMID 27477855
Abstract
Cryptococcosis, caused by Cryptococcus gattii sensu lato, is an emerging disease that was initially found in (sub)tropical regions but recently expanded to temperate regions. Cryptococcus gattii s.l. infections are mostly encountered in healthy individuals, frequently affecting both lungs and the central nervous system (CNS). Usually, C. gattii s.l. is less susceptible to antifungal compounds than its counterpart, C. neoformans s.l. We studied 18 clinical C. gattii s.l. isolates with amplified fragment length polymorphism (AFLP) fingerprinting, mating-typing, multi-locus sequence typing (MLST) and antifungal susceptibility testing. All isolates were C. deuterogattii (genotype AFLP6/VGII), 14 were mating-type α and four were type a. Amphotericin B, itraconazole, voriconazole, posaconazole and isavuconazole showed high activity, with minimum inhibitory concentration (MIC) ranges of 0.063-0.25, 0.031-0.25, 0.031-0.25, 0.031-0.25 and <0.016-0.25 μg mL
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32103
Posaconazole, VETRANAL, analytical standard
C37H42F2N8O4