EMAIL THIS PAGE TO A FRIEND
Asian-Australasian journal of animal sciences
Effects of β-Glucan on the Release of Nitric Oxide by Macrophages Stimulated with Lipopolysaccharide.
PMID 27488844
Abstract
This research analyzed the effect of β-glucan that is expected to alleviate the production of the inflammatory mediator in macrophagocytes, which are processed by the lipopolysaccharide (LPS) of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8293
Protoporphyrin IX, ≥95%
C34H34N4O4