EMAIL THIS PAGE TO A FRIEND
Journal of immunotoxicology
Chronic exposure to endosulfan induces inflammation in murine colon via β-catenin expression and IL-6 production.
PMID 27494533
Abstract
Endosulfan (ENDO) is a widely used organochlorine (OC) pesticide and persistent organo-pollutant. Epidemiological studies have shown that high levels of OC exposure were related to colorectal cancer (CRC) incidence. The objectives of the present study were to evaluate histological changes in the colon, as well as in in situ expression of β-catenin and P-selectin, and serum levels of select pro-inflammatory cytokines in mice administered ENDO; there is a relationship between increased serum IL-6 and P-selectin levels in CRC patients and aberrant β-catenin signaling is important in initiation/maintenance of most CRCs. Mice were exposed to ENDO (at dose < LD
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D161403
5,5-Dimethylhydantoin, 97%
C5H8N2O2