EMAIL THIS PAGE TO A FRIEND
Organic letters
Development of a Method for the N-Arylation of Amino Acid Esters with Aryl Triflates.
PMID 27498618
Abstract
A general method for the N-arylation of amino acid esters with aryl triflates is described. Both α- and β-amino acid esters, including methyl, tert-butyl, and benzyl esters, are viable substrates. Reaction optimization was carried out by design of experiment (DOE) analysis using JMP software. The mild reaction conditions, which use t-BuBrettPhos Pd G3 or G4 precatalyst, result in minimal racemization of the amino acid ester. This method is the first synthetic application of the t-BuBrettPhos Pd G4 precatalyst. Mechanistic studies show that the observed erosion in enantiomeric excess is due to racemization of the amino acid ester starting material and not of the product.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
900340
rac-BINAP Pd G4
C58H47NO3P2PdS
745979
tBuBrettPhos Pd G3, 96%
C44H62NO5PPdS
900329
XantPhos Pd G4
C53H47NO4P2PdS