EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Gastrointestinal and liver physiology
The kielin/chordin-like protein KCP attenuates nonalcoholic fatty liver disease in mice.
PMID 27514479
Abstract
Nonalcoholic fatty liver disease (NAFLD) is a common cause of chronic liver disease and is increasing with the rising rate of obesity in the developed world. Signaling pathways known to influence the rate of lipid deposition in liver, known as hepatic steatosis, include the transforming growth factor (TGF) superfamily, which function through the SMAD second messengers. The kielin/chordin-like protein (KCP) is a large secreted protein that can enhance bone morphogenetic protein signaling while suppressing TGF-β signaling in cells and in genetically modified mice. In this report, we show that aging KCP mutant (Kcp
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0278 RIPA Buffer