EMAIL THIS PAGE TO A FRIEND
PDA journal of pharmaceutical science and technology
Session 1.1: Viral Clearance Using Traditional, Well-Understood Unit Operations: Low pH and Detergent Viral Inactivation.
PMID 27516488