EMAIL THIS PAGE TO A FRIEND
Lipids
Olive Oil-Based Lipid Emulsions Do Not Influence Platelet Receptor Expression in Comparison to Medium and Long Chain Triglycerides In vitro.
PMID 27518386
Abstract
Lipid emulsions influence platelet aggregation and receptor expression. However, the effect on platelet function is not fully explained. Therefore, the aim of this study was to examine the influence of the lipids Lipofundin
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1573
Thrombin Receptor Activator Peptide 6, >95% (HPLC)
C34H56N10O9