EMAIL THIS PAGE TO A FRIEND
Clinical science (London, England : 1979)
Antibodies to paraoxonase 1 are associated with oxidant status and endothelial activation in rheumatoid arthritis.
PMID 27520507
Abstract
Traditional and non-traditional cardiovascular (CV) risk factors underlie CV disease occurrence in rheumatoid arthritis (RA). Recently, a functional impairment of high-density lipoprotein (HDL) has been observed. Although the actual players are unknown, anti-HDLs were associated with altered lipid profile, decreased paraoxonase 1 (PON1) activity and CV disease in RA. Therefore, we aimed to evaluate whether the presence of antibodies against PON1 may be involved in this scenario. IgG anti-PON1 antibodies were quantified by ELISA in serum samples from 212 RA patients, 175 healthy controls (HC) and 54 subjects with traditional CV risk factors (CVR). A subgroup of 13 RA patients was prospectively followed upon tumour necrosis factor-α  (TNFα) blockade. Serum PON1 activity, nitric oxide (NO) and total antioxidant capacity (TAC) were measured. Interferon-γ (IFNγ), interleukin 8 (IL-8), monocyte chemotactic protein 1 (MCP-1), vascular endothelial growth factor (VEGF), soluble intercellular adhesion molecule (sICAM) and TNFα serum levels were assessed by immunoassays. PON1 rs662 (Q > R) status was studied by reverse transcription (RT)-PCR. IgG anti-PON1 antibodies are increased in RA patients compared with HC (P<0.0001) and CVR subjects (P<0.001), even after correcting for total IgG levels. Although no associations with lipid profile were found, a positive correlation with Health Assessment Questionnaire (HAQ) was observed (r=0.215, P=0.004). Anti-PON1 antibodies were associated with PON1 activity, NO and TAC, a rs662-mediated gene-dosage effect being found. Similarly, anti-PON1 antibodies were associated with sICAM serum levels in univariate and multivariate models. Finally, these antibodies were not affected by TNFα blockade. Anti-PON1 antibodies can be responsible for PON1 impairment in RA patients, with a potential impact on biomarkers of oxidative status and endothelial activation. A gene-environment interaction of rs662 variants is supported.