EMAIL THIS PAGE TO A FRIEND
Chemosphere
Simultaneous determination of 20 trace organic chemicals in waters by solid-phase extraction (SPE) with triple-quadrupole mass spectrometer (QqQ-MS) and hybrid quadrupole Orbitrap high resolution MS (Q-Orbitrap-HRMS).
PMID 27522181
Abstract
A sensitive method for simultaneous determination of 20 trace organic chemicals (TOrCs, including preservatives, antioxidants, disinfectants, oestrogens, alkyl-phenols and bisphenol-A) in surface water and wastewater has been developed and validated based on the optimisation of solid-phase extraction (SPE) followed by liquid chromatography-mass spectrometry (LC-MS) analysis. 500xa0mL acidified (pHxa0=xa02.5) water samples were pre-concentrated by Supel-Select HLB cartridge (200xa0mg, 6xa0mL) and eluted with 12xa0mL mixture of acetonitrile and ethyl acetate (50:50, v/v). This optimised SPE procedure could provide >75% recoveries for the majority of TOrCs. The instrumental methods were developed using two different LC-MS systems: a triple-quadrupole MS (QqQ-MS) and a hybrid quadrupole Orbitrap high resolution MS (Q-Orbitrap-HRMS). Both showed good performance data, but the former system provided better linearity and method precision, with the latter system providing 2-33 times lower detection limits. Different matrix effects were observed for both systems: No remarkable matrix effects were observed for Q-Orbitrap-HRMS but significant matrix effects were found in influent and river water samples for the QqQ-MS. This analytical method was subsequently employed to analyse the TOrCs in river waters and wastewaters from China successfully, which confirmed its applicability to environmental samples.