EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
Biocompatibility of plasma-treated poly(3-hydroxybutyrate-co-3-hydroxyvalerate) nanofiber mats modified by silk fibroin for bone tissue regeneration.
PMID 27524087
Abstract
The objective of this study was to produce biocompatible plasma-treated and silk-fibroin (SF) modified poly(3-hydroxybutyrate-co-3-hydroxyvalerate) (PHBV) nanofiber mats. The mats were plasma-treated using O2 or N2 gas to increase their hydrophilicity followed by SF immobilization for the improvement of biocompatibility. Contact angle measurements and SEM showed increased hydrophilicity and no disturbed morphology, respectively. Cell proliferation assay revealed that SF modification together with N2 plasma (PS/N2) promoted higher osteoblastic (SaOs-2) cell viability. Although, O2 plasma triggered more mineral formation on the mats, it showed poor cell viability. Consequently, the PS/N2 nanofiber mats would be a potential candidate for bone tissue engineering applications.